missing translation for 'onlineSavingsMsg'
Learn More

Benperidol EP Impurity A, Mikromol™

Product Code. 18071090
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18071090 100 mg 100mg
18091610 25 mg 25mg
30463650 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18071090 Supplier Mikromol Supplier No. MM0113.03
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benperidol EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Piperidin-4-yl)-1,3-dihydro-2H-benzimidazol-2-one
CAS 20662-53-7
Analyte or Component Names 1-(Piperidin-4-yl)-1,3-dihydro-2H-benzimidazol-2-one
Shipping Condition Room Temperature
Molecular Formula C12 H15 N3 O
Certifications/Compliance ISO 9001
Synonym Benperidol Imp. A (EP), 1-(Piperidin-4-yl)-1,3-dihydro-2H-benzimidazol-2-one, Benperidol Impurity A
InChI Formula InChI=1S/C12H15N3O/c16-12-14-10-3-1-2-4-11(10)15(12)9-5-7-13-8-6-9/h1-4,9,13H,5-8H2,(H,14,16)
SMILES O=C1Nc2ccccc2N1C3CCNCC3
IUPAC Name 3-piperidin-4-yl-1H-benzimidazol-2-one
Molecular Weight (g/mol) 217.27
Quantity 100 mg
Formula Weight 217.1215 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.