missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzaldoxime, predominantly (E)-isomer, 98%

Product Code. 11459932
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459932 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459932 Supplier Alfa Aesar™ Supplier No. A12053.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Oximes are used as a peel-preventing additive in paints and lacquers. It acts as an antioxidant against oxidative drying materials which forms sticky skin with air oxygen. Oximes are used as chemical building block for the synthesis of agrochemicals and pharmaceuticals. In medicine application, Oxime structure is effective in cholinesterase reactivators to treat the poisoning by organophosphates. Oxime is used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

Solubility
Slightly soluble in water. Soluble in ethanol and ether.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases, acids, acid anhydrides and acid chlorides.
TRUSTED_SUSTAINABILITY

Specifications

CAS 932-90-1
Melting Point 33°C to 35°C
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point 108°C (226°F)
Molecular Formula C7H7NO
MDL Number MFCD00002119 MFCD00063017
Quantity 25 g
Beilstein 774138
Synonym benzaldehyde oxime, benzaldoxime, benzaldehyde, oxime, e-benzaldehyde oxime, alpha-benzaldoxime, benzaldehydeoxime, syn-benzaldoxime, benzaldehyde, oxime, e, e-n-phenylmethylidene hydroxylamine, z-benzaldoxime
InChI Key VTWKXBJHBHYJBI-SOFGYWHQSA-N
SMILES O\N=C\C1=CC=CC=C1
IUPAC Name (E)-N-(phenylmethylidene)hydroxylamine
Molecular Weight (g/mol) 121.14
PubChem CID 5324611
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material Benzaldoxime, predominantly (E)-isomer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.