missing translation for 'onlineSavingsMsg'
Learn More

Benzo[a]pyrene-4,5-quinone, TRC

Product Code. 30134199
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134199 10 mg 10mg
30011960 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134199 Supplier Toronto Research Chemicals Supplier No. TRCB20588510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42286-46-4
Molecular Formula C20H10O2
Molecular Weight (g/mol) 282.29
Synonym 4,5-Benzo[a]pyrenequinone, Benzo[pqr]tetraphene-4,5-dione;
IUPAC Name benzo[a]pyrene-4,5-dione
SMILES O=C1C(=O)c2cc3ccccc3c4ccc5cccc1c5c24

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H10O2/c21-19-15-7-3-5-11-8-9-14-13-6-2-1-4-12(13)10-16(20(19)22)18(14)17(11)15/h1-10H
Formula Weight 282.0681
Chemical Name or Material Benzo[a]pyrene-4,5-quinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.