Learn More
Benzo[b]thiophene-2-carbonyl chloride, 98%, Thermo Scientific™
Product Code. 11318005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318005 1 g 1g
11328005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11318005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08140.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39827-11-7
Molecular Formula C9H5ClOS
Molecular Weight (g/mol) 196.648
MDL Number MFCD00051635
InChI Key DNGLRCHMGDDHNC-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carbonyl chloride, thianaphthene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, acmc-20alq1, 2-benzthiophenecarbonylchloride, 2-benzothiophenecarbonyl chloride, benzothiophene-2-carbonyl chloride, 2-benzo b thiophenecarbonyl chloride, 2-chlorocarbonyl-1-benzothiophene
PubChem CID 142384
IUPAC Name 1-benzothiophene-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)C=C(S2)C(=O)Cl
Specifications
Melting Point 85°C to 88°C
Quantity 1 g
UN Number UN3261
Beilstein 125174
Sensitivity Moisture sensitive
Formula Weight 196.65
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-carbonyl chloride