missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carbonyl chloride, 98%, Thermo Scientific™

Product Code. 11328005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328005 5 g 5g
11318005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08140.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39827-11-7
Molecular Formula C9H5ClOS
Molecular Weight (g/mol) 196.648
MDL Number MFCD00051635
InChI Key DNGLRCHMGDDHNC-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carbonyl chloride, thianaphthene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, acmc-20alq1, 2-benzthiophenecarbonylchloride, 2-benzothiophenecarbonyl chloride, benzothiophene-2-carbonyl chloride, 2-benzo b thiophenecarbonyl chloride, 2-chlorocarbonyl-1-benzothiophene
PubChem CID 142384
IUPAC Name 1-benzothiophene-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)C=C(S2)C(=O)Cl

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
UN Number UN3261
Beilstein 125174
Sensitivity Moisture sensitive
Formula Weight 196.65
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.