missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzonitrile-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Benzonitrile-D5 |
| CAS | 2102-15-0 |
| Molecular Formula | C7 D5 N |
| Quantity | 0.5 g |
| Synonym | Benzonitrile-2,3,4,5,6-d5 (ACI), Benzonitrile-d5 (7 CI, 8 CI, 9 CI), Cyanobenzene-d5, Benzonitrile-D5 |
| InChI Formula | InChI=1 S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C#N)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuteriobenzonitrile |
| Molecular Weight (g/mol) | 108.152 |
| Formula Weight | 108.074 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction