missing translation for 'onlineSavingsMsg'
Learn More

Benzyl [1-Cyano-1-methylethyl]carbamate, TRC

Product Code. 30343926
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343926 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343926 Supplier Toronto Research Chemicals Supplier No. TRCB2338702.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl [1-Cyano-1-methylethyl]carbamate
CAS 100134-82-5
Molecular Formula C12 H14 N2 O2
Quantity 2.5 g
Synonym Carbamic acid, N-(1-cyano-1-methylethyl)-, phenylmethyl ester, Carbamic acid, (1-cyano-1-methylethyl)-, benzyl ester (6CI), Carbamic acid, (1-cyano-1-methylethyl)-, phenylmethyl ester (9CI), Phenylmethyl N-(1-cyano-1-methylethyl)carbamate, (Cyanodimethylmethyl)carbamic Acid Benzyl Ester, Benzyl [1-cyano-1-methylethyl]carbamate, Benzyl N-(1-cyano-1-methylethyl)carbamate
InChI Formula InChI=1S/C12H14N2O2/c1-12(2,9-13)14-11(15)16-8-10-6-4-3-5-7-10/h3-7H,8H2,1-2H3,(H,14,15)
SMILES CC(C)(NC(=O)OCc1ccccc1)C#N
IUPAC Name benzyl N-(2-cyanopropan-2-yl)carbamate
Molecular Weight (g/mol) 218.25
Formula Weight 218.1055
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.