missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,2,2-Trichloroacetimidate, TRC

Product Code. 30331771
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331771 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331771 Supplier Toronto Research Chemicals Supplier No. TRCB31607510G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 2,2,2-Trichloroacetimidate
CAS 81927-55-1
Molecular Formula C9H8Cl3NO
Quantity 10 g
Synonym 2,2,2-Trichloroethanimidic Acid Phenylmethyl Ester, 2,2,2-Trichloroacetimidic Acid Benzyl Ester, Benzyl Trichloroacetimidate, O-Benzyl 2,2,2-Trichloroacetimidate
InChI Formula InChI=1S/C9H8Cl3NO/c10-9(11,12)8(13)14-6-7-4-2-1-3-5-7/h1-5,13H,6H2
SMILES ClC(Cl)(Cl)C(=N)OCc1ccccc1
IUPAC Name benzyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol) 252.52
Formula Weight 250.9671
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.