missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5 trans-Cinnamate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | Cinnamic Acid Benzyl Ester-D5 (benzyl-2,3,4,5,6-D5) |
| CAS | 347840-02-2 |
| Molecular Formula | C16 D5 H9 O2 |
| Quantity | 0.5 g |
| Synonym | 2-Propenoic acid, 3-phenyl-, phenyl-d5-methyl ester (9 CI), 3-Phenyl-2-propenoic acid phenyl-d5-methyl ester, Benzyl-d5 cinnamate, Cinnamic acid benzyl ester-d5, Benzyl-2,3,4,5,6-d5 Cinnamate, Cinnamic acid benzyl ester-D5 (benzyl-2,3,4,5,6-D5) |
| InChI Formula | InChI=1 S/C16H14O2/c17-16(12-11-14-7-3-1-4-8-14)18-13-15-9-5-2-6-10-15/h1-12 H,13H2/i2D,5 D,6 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(COC(=O)C=Cc2ccccc2)c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)methyl 3-phenylprop-2-enoate |
| Molecular Weight (g/mol) | 243.312 |
| Formula Weight | 243.1308 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction