missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acetoacetate, TRC

Product Code. 30345900
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345900 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345900 Supplier Toronto Research Chemicals Supplier No. TRCB2100201G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Benzyl Acetoacetate
CAS 5396-89-4
Molecular Formula C11H12O3
Quantity 1 g
Synonym 3-Oxobutanoic Acid Benzyl Ester, 3-Oxobutanoic Acid Phenylmethyl Ester, 3-Oxobutyric Acid Benzyl Ester, Benzyl 3-Oxobutanoate, Benzyl 3-Oxobutyrate, Benzyl Acetoacetate, Benzyl Acetylacetate, NSC 4391, Phenylmethyl 3-Oxobutanoate
InChI Formula InChI=1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3
SMILES CC(=O)CC(=O)OCc1ccccc1
IUPAC Name benzyl 3-oxobutanoate
Molecular Weight (g/mol) 192.21
Formula Weight 192.0786
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.