missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acetoacetate, TRC

Product Code. 30331558
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331558 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331558 Supplier Toronto Research Chemicals Supplier No. TRCB210020500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Acetoacetate
CAS 5396-89-4
Molecular Formula C11H12O3
Quantity 500 mg
Synonym 3-Oxobutanoic Acid Benzyl Ester, 3-Oxobutanoic Acid Phenylmethyl Ester, 3-Oxobutyric Acid Benzyl Ester, Benzyl 3-Oxobutanoate, Benzyl 3-Oxobutyrate, Benzyl Acetoacetate, Benzyl Acetylacetate, NSC 4391, Phenylmethyl 3-Oxobutanoate
InChI Formula InChI=1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3
SMILES CC(=O)CC(=O)OCc1ccccc1
IUPAC Name benzyl 3-oxobutanoate
Molecular Weight (g/mol) 192.21
Formula Weight 192.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.