missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-alpha,alpha-d2 Bromide, CDN
Click to view available options
Quantity:
0.25 g
Unit Size:
0.25g
Specifications
Specifications
| Chemical Name or Material | Benzyl Bromide-alpha,alpha-D2 |
| CAS | 51271-29-5 |
| Molecular Formula | C7 D2 H5 Br |
| Quantity | 0.25 g |
| Synonym | Benzene, (bromomethyl-d2)- (9 CI, ACI), (Bromomethyl-d2)benzene (ACI), Toluene-α,α-d2, α-bromo- (6 CI), [Bromo(dideuterio)methyl]benzene, Benzyl bromide-α,α-D2, α,α-Dideuterobenzyl bromide, Benzyl-α,α-d2 bromide, Benzyl bromide-d2 |
| InChI Formula | InChI=1 S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5 H,6H2/i6D2 |
| SMILES | [2 H]C([2 H])(Br)c1ccccc1 |
| IUPAC Name | [bromo(dideuterio)methyl]benzene |
| Molecular Weight (g/mol) | 173.0468 |
| Formula Weight | 171.9857 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction