Learn More
Benzyl cyanide, 99+%
Product Code. 10574121 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
Product Code. 10574121
Brand: Thermo Scientific Acros undefined
Request Bulk or Custom Format
CAS: 140-29-4 | C8H7N | 117.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140-29-4
Molecular Weight (g/mol) 117.15
Specifications
Melting Point -24.0°C
Density 1.0100 g/mL
Boiling Point 233.0°C to 234.0°C
Flash Point 102°C
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C8H7N
Linear Formula C6H5CH2CN
Refractive Index 1.5220 to 1.5240
Quantity 500 mL
Beilstein 09, 441
Merck Index 15, 1134
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 117.15
Chemical Name or Material Benzyl cyanide
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302-if on skin: .
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P352-Wash with plenty of water/ .
Supplemental information
  • MIXTURE LIST-Contains : Benzyl cyanide