missing translation for 'onlineSavingsMsg'
Learn More

Benzyl nicotinate, 98%

Product Code. 11395189
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395189 50 g 50g
11305199 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395189 Supplier Thermo Scientific Alfa Aesar Supplier No. A18505.18
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 94-44-0 | C13H11NO2 | 213.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-44-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00023584
InChI Key KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Synonym benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
PubChem CID 7191
ChEBI CHEBI:31268
IUPAC Name benzyl pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2

Specifications

Melting Point 21°C to 23°C
Density 1.165
Boiling Point 179°C to 180°C (5 mmHg)
Flash Point 170°C (338°F)
Refractive Index 1.57
Quantity 50 g
Beilstein 159169
Merck Index 14,6526
Formula Weight 213.24
Percent Purity 99%
Chemical Name or Material Benzyl nicotinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.