missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Nicotinate, Mikromol™

Product Code. 17932216
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932216 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932216 Supplier Mikromol Supplier No. MM0104.00
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyl Nicotinate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinic acid benzyl ester
CAS 94-44-0
Analyte or Component Names Nicotinic acid benzyl ester
Shipping Condition Room Temperature
Molecular Formula C13 H11 N O2
Certifications/Compliance ISO 17025
Synonym Benzyl Nicotinate, 3-Pyridinecarboxylic acid phenylmethyl ester, Nicotinic acid benzyl ester, Benzyl nicotinate, Benzyl pyridine-3-carboxylate, Niacin benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Rubriment
InChI Formula InChI=1S/C13H11NO2/c15-13(12-7-4-8-14-9-12)16-10-11-5-2-1-3-6-11/h1-9H,10H2
SMILES O=C(OCc1ccccc1)c2cccnc2
IUPAC Name benzyl pyridine-3-carboxylate
Molecular Weight (g/mol) 213.23
Quantity 500 mg
Formula Weight 213.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.