missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Nicotinate, TRC

Product Code. 30329376
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329376 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329376 Supplier Toronto Research Chemicals Supplier No. TRCB285465100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinic acid benzyl ester
CAS 94-44-0
Molecular Formula C13 H11 N O2
Quantity 100 g
Synonym Benzyl Nicotinate, 3-Pyridinecarboxylic acid phenylmethyl ester, Nicotinic acid benzyl ester, Benzyl nicotinate, Benzyl pyridine-3-carboxylate, Niacin benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Rubriment
InChI Formula InChI=1S/C13H11NO2/c15-13(12-7-4-8-14-9-12)16-10-11-5-2-1-3-6-11/h1-9H,10H2
SMILES O=C(OCc1ccccc1)c2cccnc2
IUPAC Name benzyl pyridine-3-carboxylate
Molecular Weight (g/mol) 213.23
Formula Weight 213.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.