missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Penicillinate-d7 Potassium Salt (Penicillin G-d7 Potassium Salt), TRC

Product Code. 17988999
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988999 10 mg 10mg
17919139 2.5 mg 2.5mg
17978279 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17988999 Supplier Toronto Research Chemicals Supplier No. TRCB28860010MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352323-25-2
Molecular Formula C16H10D7KN2O4S
Molecular Weight (g/mol) 379.52
Synonym Penicillin G Potassium D7 (Benzyl D7)
IUPAC Name potassium;(2S,5R,6R)-6-[[2,2-dideuterio-2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1/i3D,4D,5D,6D,7D,8D2;
Formula Weight 379.0985
Percent Purity >95%
Chemical Name or Material Benzylpenicillinate-d7, Potassium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.