missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylmethyl-d3-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-Methylbenzylamine-d3 |
| CAS | 122025-09-6 |
| Molecular Formula | C8 D3 H8 N |
| Quantity | 0.1 g |
| Synonym | Benzenemethanamine, N-(methyl-d3)- (9 CI), N-(Methyl-d3)benzenemethanamine, N-(Methyl-d3)benzylamine, N-Methylbenzylamine-D3 (N-methyl-D3), N-Methylbenzylamine-d3, Benzylmethyl-d3-amine |
| InChI Formula | InChI=1 S/C8H11N/c1-9-7-8-5-3-2-4-6-8/h2-6,9 H,7H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])NCc1ccccc1 |
| IUPAC Name | N-benzyl-1,1,1-trideuteriomethanamine |
| Molecular Weight (g/mol) | 124.198 |
| Formula Weight | 124.108 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction