missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetic acid, 95%

Product Code. 15460838
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460838 Supplier Thermo Scientific Alfa Aesar Supplier No. H54581.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 30379-55-6 | C9H10O3 | 166.18 g/mol

It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterification, tetra aqua(1,10-phenanthroline-[κ]2N,N?)magnesium(II) bis[(2,4-dichlorophenyl)acetate], as ligand in the preparation of diaquabis(benzyloxyacetato)copper(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterific
TRUSTED_SUSTAINABILITY

Specifications

CAS 30379-55-6
Density 1.162
Boiling Point 137°C to 139°C
Flash Point 113°C (235°F)
Molecular Formula C9H10O3
Linear Formula C6H5CH2OCH2COOH
Refractive Index 1.526
MDL Number MFCD00274211
Quantity 5 g
Synonym benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy
InChI Key GRZHHTYDZVRPIC-UHFFFAOYSA-N
SMILES OC(=O)COCC1=CC=CC=C1
IUPAC Name 2-phenylmethoxyacetic acid
Molecular Weight (g/mol) 166.18
PubChem CID 290301
Formula Weight 166.17
Percent Purity 95%
Chemical Name or Material Benzyloxyacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.