missing translation for 'onlineSavingsMsg'
Learn More

Berberrubine Chloride, TRC

Product Code. 30009914
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009914 1 mg 1mg
30005392 2.5 mg 2.5mg
30011910 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009914 Supplier Toronto Research Chemicals Supplier No. TRCB2000951MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15401-69-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
Synonym 9-Berberoline Chloride, 9-Demethoxy-9-hydroxyberberinium Chloride, Berberrubine Chloride, Beroline Chloride;
IUPAC Name 17-methoxy-5,7-dioxa-13-azoniapentacyclo[11.8.0.02,10.04,8.015,20]henicosa-1(13),2,4(8),9,14,16,18,20-octaen-16-ol;chloride
SMILES [Cl-].COc1ccc2cc3c4cc5OCOc5cc4CC[n+]3cc2c1O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H15NO4.ClH/c1-22-16-3-2-11-6-15-13-8-18-17(23-10-24-18)7-12(13)4-5-20(15)9-14(11)19(16)21;/h2-3,6-9H,4-5,10H2,1H3;1H
Formula Weight 357.0768
Chemical Name or Material Berberrubine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.