Learn More
Emma Safety Footwear Berlin Safety Shoes
Woven textile combined with PU provides a wear-resistant polyurethane that optimally protects the textile. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot. Thanks to the Hydro-Tec™ lining, this safety shoe is also particularly breathable.
Brand: Emma Safety Footwear MM40664744
Additional Details : Weight : 1.66800kg
Description
- S1P-Safety shoe with sole plate for dry work
- Woven Textile Polyurethane coated construction
- Metal-free
- Chrome-free lining
- Hydro-Tec™ Antibacterial lining
- ESD certified
Specifications
EN ISO 20345:2011 | |
Gray, Blue | |
Hydro-Tec™ Antibacterial | |
44 |
Laces | |
Berlin | |
Textile | |
Composite |