missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard Solution (Spectro ECON), Chem Lab™

Product Code. 11979022
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11979022 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11979022 Supplier Chem-Lab Supplier No. CL01.0217.0500
  • ONLINE EXCLUSIVE PRICE
    52.00€
    25.50€ / 500mL
    Save 26.50€
    50% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Concentration 1000 μg/mL
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • Beryllium standard solution 1000 µg/ml
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.