missing translation for 'onlineSavingsMsg'
Learn More

Besifloxacin Hydrochloride (Contains ~15% Inorganics), TRC

Product Code. 30138595
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138595 50 mg 50mg
30137183 25 mg 25mg
30005739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138595 Supplier Toronto Research Chemicals Supplier No. TRCB31900050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405165-61-9
Molecular Formula C19 H21 Cl F N3 O3 . Cl H
Molecular Weight (g/mol) 430.3
Synonym 3-Quinolinecarboxylic acid, 7-[(3R)-3-aminohexahydro-1H-azepin-1-yl]-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, hydrochloride (1:1), 3-Quinolinecarboxylic acid, 7-[(3R)-3-aminohexahydro-1H-azepin-1-yl]-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, monohydrochloride (9CI), BOL 303224A, Besifloxacin hydrochloride, Besivance, SS 734
IUPAC Name 7-[(3R)-3-aminoazepan-1-yl]-8-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES Cl.N[C@@H]1CCCCN(C1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2Cl)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H21ClFN3O3.ClH/c20-15-16-12(18(25)13(19(26)27)9-24(16)11-4-5-11)7-14(21)17(15)23-6-2-1-3-10(22)8-23;/h7,9-11H,1-6,8,22H2,(H,26,27);1H/t10-;/m1./s1
Formula Weight 429.1022
Chemical Name or Material Besifloxacin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.