Learn More
beta 2-Microglobulin Antibody (B2M/1118), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47702FR
Product Code. 18764213
  • 588.00€ / 0.10mL
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Cell Surface), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta 2-Microglobulin
Monoclonal
DyLight 680
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Full length recombinant human beta 2-Microglobulin protein (Uniprot: P61769)
0.1 mL
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
567
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
B2M/1118
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Primate