missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Amyloid Antibody (MOAB-2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-13075CL1
This item is not returnable.
View return policy
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Bacteria,Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunoblot,Dot Blot,Immunohistochemistry Free-Floating,Immunocytochemistry/Immunofluorescence
Specifications
| beta Amyloid | |
| Monoclonal | |
| CoraFluor 1 | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| This beta Amyloid antibody was developed against recombinant human beta Amyloid 42. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 351 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| MOAB-2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bacteria, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction