missing translation for 'onlineSavingsMsg'
Learn More

Beta-Caryophyllene, TRC

Product Code. 17934935
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934935 1 mL 1mL
30003356 25 mL 25mL
17964055 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17934935 Supplier Toronto Research Chemicals Supplier No. TRCC1847251ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-44-5
Molecular Formula C15 H24
Molecular Weight (g/mol) 204.35
Synonym Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, (1R,4E,9S)-, Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, (E)-(1R,9S)-(-)- (8CI), Bicyclo[7.2.0]undec-4-ene, 4,11,11-trimethyl-8-methylene-, [1R-(1R*,4E,9S*)]-, (-)-(E)-Caryophyllene, (-)-Caryophyllene, (-)-E-Caryophyllene, (-)-trans-Caryophyllene, (-)-β-Caryophyllene, (1R,4E,9S)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (E)-Caryophyllene, Caryophyllene, Caryophyllene B, NSC 11906, l-Caryophyllene, trans-Caryophyllene, trans-β-Caryophyllene, β-Caryophyllen, β-Caryophyllene, β-Caryophyllene, (-)
IUPAC Name (1R,4E,9S)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene
SMILES C\C\1=C/CCC(=C)[C@H]2CC(C)(C)[C@@H]2CC1

Specifications

Quantity 1 mL
InChI Formula InChI=1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6+/t13-,14-/m1/s1
Formula Weight 204.1878
Percent Purity >95%
Chemical Name or Material (-)-Trans-Caryophyllene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.