missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Maltose octaacetate, 98%

Product Code. 11479863
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479863 25 g 25g
11459863 1 g 1g
11469863 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479863 Supplier Thermo Scientific Alfa Aesar Supplier No. A12988.14
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 22352-19-8 | C28H38O19 | 678.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22352-19-8
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.593
MDL Number MFCD00079432
InChI Key WOTQVEKSRLZRSX-FKNZMNJBSA-N
Synonym Octa-O-acetyl-beta-D-maltose
PubChem CID 126963554
IUPAC Name [(2S,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 157°C to 160°C
Quantity 25 g
Beilstein 79279
Optical Rotation +65° (c=10 in Chloroform)
Formula Weight 678.6
Percent Purity 98%
Chemical Name or Material beta-D-Maltose octaacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.