missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (DC1 4C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-2619-0.025mg
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| beta Galactosidase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Mouse | |
| 130 kDa | |
| 0.025 mg | |
| Beta Galactosidase Epitope Tags, Epitope Tags, Fusion Proteins | |
| Beta-galactosidase (DC1 4C7). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| DC1 4C7 | |
| Unconjugated | |
| EC 3.2.1.23; b-gal | |
| Beta-galactosidase EC.3.2.1.23 from E.coli | |
| Protein A purified | |
| RUO | |
| Primary | |
| E. coli | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction