Learn More
beta-III Tubulin Antibody (CL5813) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61430AF488
Product Code. 30298411
  • 737.00€ / 0.10mL
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-III Tubulin
Monoclonal
Alexa Fluor 488
beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
This antibody was developed using a synthetic peptide derived from Q13509, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific
10381
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5813
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat