missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (CL5814) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61431JF525
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| Janelia Fluor 525 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| This antibody was developed using a synthetic peptide derived from Q13509, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
| 10381 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5814 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction