Learn More
Invitrogen™ Beta Lactoglobulin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119523
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Beta-lactoglobulin is a major protein found in dairy products and is not present in human milk. It has been identified as a potential milk allergen, and is responsible for transport of both iron and hydrophobic molecules.
Specifications
| Beta Lactoglobulin | |
| Polyclonal | |
| Unconjugated | |
| PAEP | |
| beta lactoglobulin C; beta lactoglobulin D; beta-lactoglobulin; Beta-LG; BLG; bLG C; Bos d 5; lactoglobulin, beta; LGB; PAEP; progestagen-associated endometrial protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 280838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
| ELISA, Functional Assay, Immunoelectrophoresis, Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P02754 | |
| PAEP | |
| Beta lactoglobulin extract from bovine milk. | |
| 1 mL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.