Learn More
beta-Propiolactone, 98%, pure
Product Code. 10741521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10741521 5 mL Glass bottle 5mL
10625511 25 mL Glass bottle 25mL
10358153 100 mL Glass bottle 100mL
11351523 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10741521 Supplier Thermo Scientific Acros Supplier No. 269040050
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
CAS: 57-57-8 | C3H4O2 | 72.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Packaging Note: Glass bottle is put into a can for shipping.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-57-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00005169
InChI Key VEZXCJBBBCKRPI-UHFFFAOYSA-N
Synonym beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
PubChem CID 2365
ChEBI CHEBI:49073
IUPAC Name oxetan-2-one
SMILES O=C1CCO1
Specifications
Density 1.1460g/mL
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 mL
Formula Weight 72.06
Grade Pure
Chemical Name or Material β-Propiolactone, Pure