missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Betacellulin Monoclonal Antibody (197415)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA560865

Product Code. 30534811

  • 589.00€ / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Betacellulin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q05928
BTC
E. coli-derived recombinant mouse Betacellulin/BTC, Asp32-Gln118.
500 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
197415
Unconjugated
BTC
Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Rat
Protein A/G
RUO
12223
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.