missing translation for 'onlineSavingsMsg'
Learn More

Betahistine, TRC

Product Code. 30179060
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179060 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179060 Supplier Toronto Research Chemicals Supplier No. TRCB324005500MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5638-76-6
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.19
Synonym N-Methyl-2-pyridineethanamine, 2-(2-Methylaminoethyl)pyridine, 2-(beta-Methylaminoethyl)pyridine, Methyl(2-pyridin-2-ylethyl)amine, N-Methyl-2-(2-pyridinyl)ethanamine, N-Methyl-2-(2-pyridyl)ethylamine, N-Methyl-2-pyridineethylamine, N-Methyl-N-[2-(2-pyridinyl)ethyl]amine, N-Methyl-N-[2-(pyridin-2-yl)ethyl]amine, N-Methyl[2-(pyridin-2-yl)ethyl]amine, N-[2-(2-Pyridyl)ethyl]-N-methylamine, NSC 42617, OBE 101, Y-G 14, [2-(2-Pyridyl)ethyl]methylamine, beta-Histine;
IUPAC Name N-methyl-2-pyridin-2-ylethanamine
SMILES CNCCc1ccccn1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H12N2/c1-9-7-5-8-4-2-3-6-10-8/h2-4,6,9H,5,7H2,1H3
Formula Weight 136.1
Chemical Name or Material Betahistine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.