missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betamethasone 21-Phosphate Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Betamethasone Sodium Phosphate |
| CAS | 151-73-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H28 F O8 P . 2 Na |
| Quantity | 100 mg |
| Synonym | Betamethasone Sodium Phosphate, Dexamethasone Sodium Phosphate Imp. B (EP), (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, Bentelan, Betamethasone 21-disodium phosphate, Betamethasone 21-phosphate disodium salt, Betamethasone disodium phosphate, Betamethasone sodium phosphate, Betnesol, Celestan solubile, Disodium betamethasone 21-phosphate, Durabetason, NSC 90616, Steronema, Vista-Methasone |
| InChI Formula | InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15-,16-,17-,19-,20-,21-,22-;;/m0../s1 |
| SMILES | [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-] |
| IUPAC Name | disodium;[2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
| Molecular Weight (g/mol) | 516.4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?