missing translation for 'onlineSavingsMsg'
Learn More

(BetaR,DeltaR)-Atorvastatin tert-Butyl Ester (>90%), TRC

Product Code. 30134660
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134660 100 mg 100mg
30006638 50 mg 50mg
30011622 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134660 Supplier Toronto Research Chemicals Supplier No. TRCA791740100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134395-00-9
Molecular Formula C37 H43 F N2 O5
Molecular Weight (g/mol) 614.75
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, 1,1-dimethylethyl ester, (βR,δR)-, 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, 1,1-dimethylethyl ester, [R-(R*,R*)]-, (3R,5R)-7-[2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-phenylcarbamoylpyrrol-1-yl]-3,5-dihydroxyheptanoic acid tert-butyl ester, Atorvastatin tert-butyl ester
IUPAC Name tert-butyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)OC(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C37H43FN2O5/c1-24(2)34-33(36(44)39-28-14-10-7-11-15-28)32(25-12-8-6-9-13-25)35(26-16-18-27(38)19-17-26)40(34)21-20-29(41)22-30(42)23-31(43)45-37(3,4)5/h6-19,24,29-30,41-42H,20-23H2,1-5H3,(H,39,44)/t29-,30-/m1/s1
Formula Weight 614.3156
Chemical Name or Material Atorvastatin tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.