missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEX4 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98166V
This item is not returnable.
View return policy
Description
BEX4 Polyclonal antibody specifically detects BEX4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BEX4 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 56271 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| BEX family member 4, BEX1-like protein 1, BEXL1, brain expressed X-linked-like 1, brain expressed, X-linked 4, Brain-expressed X-linked protein 4, FLJ10097, NADE3, Nerve growth factor receptor-associated protein 3, protein BEX4 | |
| Produced in rabbits immunized with E. coli-derived Human BEX4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur