missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate EP Impurity D, Mikromol™

Product Code. 30052113
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052113 25 mg 25mg
30049086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052113 Supplier Mikromol Supplier No. MM0063.020025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bezafibrate EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
CAS 41859-58-9
Analyte or Component Names Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Shipping Condition Room Temperature
Molecular Formula C21 H24 Cl N O4
Certifications/Compliance ISO 17025
Synonym Bezafibrate Imp. D (EP), Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Bezafibrate Impurity D
InChI Formula InChI=1S/C21H24ClNO4/c1-4-26-20(25)21(2,3)27-18-11-5-15(6-12-18)13-14-23-19(24)16-7-9-17(22)10-8-16/h5-12H,4,13-14H2,1-3H3,(H,23,24)
SMILES CCOC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1
IUPAC Name ethyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Molecular Weight (g/mol) 389.87
Quantity 25 mg
Formula Weight 389.1394 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.