missing translation for 'onlineSavingsMsg'
Learn More

Bezafibrate Ethyl Ester, TRC

Product Code. 30006011
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006011 25 mg 25mg
30005430 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006011 Supplier Toronto Research Chemicals Supplier No. TRCB34101025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41859-58-9
Molecular Formula C21 H24 Cl N O4
Molecular Weight (g/mol) 389.87
Synonym Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Bezafibrate Imp. D (EP)
IUPAC Name ethyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H24ClNO4/c1-4-26-20(25)21(2,3)27-18-11-5-15(6-12-18)13-14-23-19(24)16-7-9-17(22)10-8-16/h5-12H,4,13-14H2,1-3H3,(H,23,24)
Formula Weight 389.1394
Chemical Name or Material Ethyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.