Learn More
BFSP2 Monoclonal Antibody (OTI2B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804359
Description
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex.Specifications
| BFSP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13515 | |
| BFSP2 | |
| Human recombit protein fragment corresponding to amino acids 184-415 of human BFSP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2B7 | |
| Unconjugated | |
| BFSP2 | |
| CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8419 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.