missing translation for 'onlineSavingsMsg'
Learn More

Bibenzonium Bromide, LoGiCal

Product Code. 30462289
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462289 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462289 Supplier LoGiCal Supplier No. LGCFOR1835.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Bibenzonium Bromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bibenzonium Bromide
CAS 15585-70-3
Analyte or Component Names Bibenzonium Bromide
Shipping Condition Room Temperature
Molecular Formula C19 H26 N O . Br
Certifications/Compliance ISO 9001
Synonym Bibenzonium Bromide, Ethanaminium, 2-(1,2-diphenylethoxy)-N,N,N-trimethyl-, bromide (1:1), Ammonium, [2-(1,2-diphenylethoxy)ethyl]trimethyl-, bromide (8 CI), Ethanaminium, 2-(1,2-diphenylethoxy)-N,N,N-trimethyl-, bromide (9 CI), [2-(1,2-Diphenylethoxy)ethyltrimethyl]ammonium bromide (6 CI), Bibenzonium bromide, ES 132, Lysbex, Lysibex, Lysobex, Medipectol, OM-Tussic, OM-Tussis, Rea-tos, Sedobex, Thoragol
InChI Formula InChI=1 S/C19H26NO.BrH/c1-20(2,3)14-15-21-19(18-12-8-5-9-13-18)16-17-10-6-4-7-11-17;/h4-13,19 H,14-16H2,1-3H3;1 H/q+1;/p-1
SMILES [Br-].C[N+](C)(C)CCOC(Cc1ccccc1)c2ccccc2
IUPAC Name 2-(1,2-diphenylethoxy)ethyl-trimethylazanium;bromide
Industry Type Forensics
Molecular Weight (g/mol) 364.32
Quantity 10 mg
Formula Weight 363.1198 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.