Learn More
R&D Systems™ BIBP 3226 trifluoroacetate
Description
BIBP 3226 trifluoroacetate is a mixed non-peptide neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist (Ki values are 1.1, 79, 108, > 1000, > 1000 and >1000 for rNPY Y1, hNPFF2, rNPFF, rNPY Y2, rNPY Y4 and rNPY Y5 respectively). Produces an anxiogenic-like effect in rats following i.c.v. administration.
Specifications
Specifications
| Chemical Name or Material | N-[(1R)]-4-[(Aminoiminomethyl)amino-1-[[[(4-hydroxyphenyl)methyl]amino]carbonyl]butyl-α-phenylbenzeneacetamide trifluoroacetate |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | NPY Receptor Antagonists |
| Molecular Formula | C27H31N5O3.xCF3CO2H |
| Purity | 98% |