missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, Tocris Bioscience™

Product Code. 18035582
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035582 10 mg 10mg
18095482 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18035582 Supplier Tocris Bioscience Supplier No. 3389/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Non-steroidal androgen receptor antagonist

Orally active non-steroidal androgen receptor antagonist (IC50 = 190 nM). Displays peripheral selectivity and does not effect serum levels of LH and testosterone. Exhibits potent anticancer activity in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90357-06-5
Molecular Formula C18H14F4N2O4S
Molecular Weight (g/mol) 430.374
InChI Key LKJPYSCBVHEWIU-UHFFFAOYSA-N
Synonym bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
PubChem CID 2375
IUPAC Name N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
SMILES CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O

Specifications

Quantity 10 mg
Formula Weight 430.37
Percent Purity >99%
Chemical Name or Material Bicalutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.