missing translation for 'onlineSavingsMsg'
Learn More

Bidimazium Iodide, LoGiCal

Product Code. 30457470
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457470 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457470 Supplier LoGiCal Supplier No. LGCFOR1836.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Bidimazium Iodide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bidimazium Iodide
CAS 21817-73-2
Analyte or Component Names Bidimazium Iodide
Shipping Condition Room Temperature
Molecular Formula C26 H25 N2 S . I
Certifications/Compliance ISO 9001
Synonym Thiazolium, 4-[1,1'-biphenyl]-4-yl-2-[2-[4-(dimethylamino)phenyl]ethenyl]-3-methyl-, iodide (1:1), Thiazolium, 4-(4-biphenylyl)-2-[p-(dimethylamino)styryl]-3-methyl-, iodide (8 CI), Thiazolium, 4-[1,1'-biphenyl]-4-yl-2-[2-[4-(dimethylamino)phenyl]ethenyl]-3-methyl-, iodide (9 CI), Bidimazium iodide
InChI Formula InChI=1 S/C26H25N2S.HI/c1-27(2)24-16-9-20(10-17-24)11-18-26-28(3)25(19-29-26)23-14-12-22(13-15-23)21-7-5-4-6-8-21;/h4-19 H,1-3H3;1 H/q+1;/p-1
SMILES [I-].CN(C)c1ccc(\C=C/c2scc(c3ccc(cc3)c4ccccc4)[n+]2 C)cc1
IUPAC Name N,N-dimethyl-4-[(Z)-2-[3-methyl-4-(4-phenylphenyl)-1,3-thiazol-3-ium-2-yl]ethenyl]aniline;iodide
Industry Type Forensics
Molecular Weight (g/mol) 524.46
Quantity 5 mg
Formula Weight 524.0783 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.