Learn More
Econix™ Bio-bins™ Orange Paper-Based Non-Sharps Infectious Clinical Waste Containers for Alternative Treatment / Incineration
GREENER_CHOICE
Product Code. 31741153 Shop All Econix Products
Change view
Click to view available options
Color:
Orange
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31741153 Orange Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31741153 Supplier Econix™ Supplier No. BEXBB50OR
  • ONLINE EXCLUSIVE PRICE
    95.98€
    74.04€ / Pack of 8
    Save 21.94€
    22% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Great alternative to plastic bins! Bio-bins™ Orange Paper Based Non-Sharps Infectious Clinical Waste Containers. Ideal for Alternative treatment / Incineration.
Use Econix™ Bio-bins™ The Bio-bin™ is a Paper based Non-sharps Container used to dispose of single use implements, tips, petri dishes, pads and other lab waste (excluding needles and blades) which reduces over filling issues for your non-sharp clinical waste. The Bio-bin has a built-in temporary seal, absorbent mat and is waterproof.
  • 96% paper based non-plastic single-use non-sharps waste container
  • Save up to 91% carbon compared to plastic alternatives
  • Comes fold-flat for easy storage
  • Rigid, leakproof construction
  • Leakproof, built-in lid
  • Built-in absorbent Mat
  • UN 3291 approved
  • Includes temporary seal
  • Autoclavable
Specifications
Capacity (Metric) 50 L
Certifications/Compliance UN3291
Closure Type Adhesive Tape
Capacity (English) 11 gal.
Color Orange
Disposable Single-use
Height (Metric) 785 mm open, 540 mm closed
Length (Metric) 310 mm
Quantity 8 Pk.
Stackable Yes, when closed
Sterility Non-sterile
Width (Metric) 310 mm
Autoclavable Autoclavable
Liner Material Recycled Cardboard
Material Paper
Product Type Non-sharps Waste Container