Learn More
Novacyt - Microgen Bioproducts Biochemical Identification System: Staph spp.
Product Code. 15997112 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
20 Tests
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15997112 20 Tests 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15997112 Supplier Novacyt - Microgen Bioproducts Supplier No. MID69CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Biochemical identification system for Staph spp. from foods and other environmental samples.
  • Twelve-substrate biochemical identification system.
  • From a range of different technologies for the specific detection of Staph spp. from foods and other environmental samples.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
For Use With (Application) Identification of Staph Spp.
Quantity 20 Tests