missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Nano 5™ Non-sterile Nitrile Gloves, ISO Class 5

Product Code. 15268268
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 15268268

Brand: Nitritex™ NAN5XL

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

With textured fingers to provide enhanced grip. Nitritex™ BioClean Nano 5™ Non-sterile Nitrile Gloves, ISO Class 5 are antistatic, powder-free, latex-free, resistant to a wide range of chemicals and are ideal for use in electrically sensitive applications.

  • Resistant to a wide range of chemicals
  • Textured fingers for enhanced grip
  • Powder-free and latex-free
  • Excellent antistatic properties
  • Non-particulating EasyTear™ packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC)2023/2006
Color White
Disposable Single-use
Exterior Finish Fully Textured
Grade Category 3 - PPE Directive 89/686/EEC
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Finger Style Textured
For Use With (Application) Electrically sensitivity applications
Glove Cuff Style Beaded
Packaging Type EasyTear™
Quantity 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Resistant To A broad range of chemicals
Series BioClean Nano 5
Length (English) 12 in.
Length (Metric) 30 cm
Width (Metric) 115 mm ± 5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.