missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Designed for biomedical waste
- Biohazard symbol in black ink
- DOT specification displayed on each bag
- Compatible with BLSSH series of Micronova Binliner™ stands
- Liners are gamma irradiated for aseptic
- Compatable with BLSSH series BinLiner Stand
- Flat sealed
- Good compatibility with common cleaners
- Gamma irradiated liners are sterile for aseptic areas
Specifications
Specifications
| Color | Red |
| Disposable | Single-use |
| Length (Metric) | 533.4 mm |
| Sterility | Sterile |
| Width (Metric) | 914.4 mm |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Binliner Stands |
| Width (English) | 36 in. |
| Length (English) | 21 in. |
| Thickness | 3.6 mil |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?