missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific Biohazard Pipet Keeper Container
Shop All Thermo Scientific Incubateurs à CO2 Products

Description
- Pipet keeper provides a reliable solution for collecting lab contaminants while keeping workstations tidy and compliant
- Sturdy paperboard construction with visible biohazard symbols
- Self-locking top and re-closable flaps for secure containment
- Easily disposable after autoclaving per regulations
- Cost-effective and user-friendly design
Specifications
Specifications
| Capacity (English) | 4200 cc |
| Color | White, Orange, Blue |
| Dimensions (L x W x H) | 8.89 x 11.43 x 41.91 cm (3.5 x 4.5 x 16.5 in.) |
| Quantity | 50/Cs. |
| Autoclavable | Autoclavable |
| Includes | Pipet Keeper |
| Material | Plastic-lined Paperboard |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?