Learn More
Biotin X, TRC
Product Code. 30336346
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336346 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336346 Supplier Toronto Research Chemicals Supplier No. TRCB3956701G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Biotinylcaproic Acid
CAS 72040-64-3
Molecular Formula C16 H27 N3 O4 S
Quantity 1 g
Synonym N-Biotinylcaproic Acid, 6-[[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]hexanoic Acid, 6-[(Biotinyl)amino]hexanoic acid, 1H-Thieno[3,4-d]imidazole, Hexanoic Acid Deriv., 6-(Biotinylamino)caproic Acid, 6-Biotinamidocaproic Acid
InChI Formula InChI=1S/C16H27N3O4S/c20-13(17-9-5-1-2-8-14(21)22)7-4-3-6-12-15-11(10-24-12)18-16(23)19-15/h11-12,15H,1-10H2,(H,17,20)(H,21,22)(H2,18,19,23)/t11-,12-,15-/m0/s1
SMILES OC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC Name 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoic acid
Molecular Weight (g/mol) 357.47
Formula Weight 357.1722